Cyclohexyl-α-MIDA-boryl aldehyde
Synonym(s):
Cyclohexyl(6-methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)acetaldehyde
Empirical Formula (Hill Notation):
C13H20BNO5
CAS Number:
Molecular Weight:
281.11
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing