![Creatine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/763/121/ca025e71-2902-48aa-b96b-3ad4a48768d7/640/ca025e71-2902-48aa-b96b-3ad4a48768d7.png)
Creatine
![Creatine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/763/121/ca025e71-2902-48aa-b96b-3ad4a48768d7/640/ca025e71-2902-48aa-b96b-3ad4a48768d7.png)
Synonym(s):
Creatine monohydrate
Empirical Formula (Hill Notation):
C4H9N3O2 · H2O
CAS Number:
Molecular Weight:
149.15
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing