
Coproporphyrin I dihydrochloride

Synonym(s):
3,8,13,18-Tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropionic acid dihydrochloride
Empirical Formula (Hill Notation):
C36H38N4O8 · 2HCl
CAS Number:
Molecular Weight:
727.63
Products
Product Category
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing