
Clindamycin hydrochloride

Synonym(s):
(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin
Empirical Formula (Hill Notation):
C18H33ClN2O5S · HCl
CAS Number:
Molecular Weight:
461.44
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing