
Citalopram Related Compound E

Synonym(s):
1-(3-Dimethylaminopropyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile N-oxide hydrochloride
Empirical Formula (Hill Notation):
C20H21FN2O2 · HCl
Molecular Weight:
376.85
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2