
Ciclesonide

Empirical Formula (Hill Notation):
C32H44O7
CAS Number:
Molecular Weight:
540.69
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing