
Chlorodi(o-tolyl)phosphine

Synonym(s):
Bis(2-methylphenyl)phosphinous chloride, Chlorobis(2-methylphenyl)phosphine
Empirical Formula (Hill Notation):
C14H14ClP
CAS Number:
Molecular Weight:
248.69
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing