![Calcium chloride dihydrate crystalline, Ph. Eur., JP, USP, FCC, E 509](/deepweb/assets/sigmaaldrich/product/structures/214/856/c7fe6bae-c3fa-4cec-83c7-757f25a6379d/640/c7fe6bae-c3fa-4cec-83c7-757f25a6379d.png)
Calcium chloride dihydrate
![Calcium chloride dihydrate crystalline, Ph. Eur., JP, USP, FCC, E 509](/deepweb/assets/sigmaaldrich/product/structures/214/856/c7fe6bae-c3fa-4cec-83c7-757f25a6379d/640/c7fe6bae-c3fa-4cec-83c7-757f25a6379d.png)
Synonym(s):
Calcium chloride dihydrate
Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-19 of 19
Compare
Product Number
Description
Pricing