
Bumetanide Related Compound B

Synonym(s):
3-Nitro-4-phenoxy-5-sulphamoylbenzoic acid
Empirical Formula (Hill Notation):
C13H10N2O7S
CAS Number:
Molecular Weight:
338.29
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2