
Bumetanide Related Compound A

Synonym(s):
3-Amino-4-phenoxy-5-sulphamoylbenzoic acid, Desbutylbumetanide
Empirical Formula (Hill Notation):
C13H12N2O5S
CAS Number:
Molecular Weight:
308.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2