
Bisphenol E

Synonym(s):
4,4′-Ethylidenebisphenol, 1,1-Bis(4-hydroxyphenyl)ethane, Bisphenol E
Linear Formula:
CH3CH(C6H4OH)2
CAS Number:
Molecular Weight:
214.26
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing