
Asparagine anhydrous

Synonym(s):
L-Asparagine, (S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide, Asparagine Anhydrous
Linear Formula:
H2NCOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
132.12
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2