
Artemether

Synonym(s):
Dihydroartemisinin methyl ether, Dihydroqinghaosu methyl ether, SM-224
Empirical Formula (Hill Notation):
C16H26O5
CAS Number:
Molecular Weight:
298.37
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Special Grade
Showing 1-3 of 3