
Anti-MTHFD1 antibody produced in rabbit

Synonym(s):
Anti-C1-THF synthase antibody produced in rabbit, Anti-C1-Tetrahydrofolate synthase, cytoplasmic antibody produced in rabbit, Anti-MTHFC, Anti-MTHFD, Anti-Methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing