Anti-JNK, Activated (Diphosphorylated JNK) antibody, Mouse monoclonal
Synonym(s):
Monoclonal Anti-JNK, Activated (Diphosphorylated JNK) antibody produced in mouse
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing