
Anti-Glutathione-S-Transferase (GST)–Peroxidase Conjugate antibody produced in rabbit

Synonym(s):
Anti-Glutathione-S-Transferase, Anti-GST
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing