Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit
Synonym(s):
Anti-c-Myc
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Analyte Suitability
Antibody Form
Research Area
Conjugate
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing