Skip to Content
Merck
Anti-CD276 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-CD276 antibody produced in rabbit

Anti-CD276 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-4Ig-B7-H3 antibody produced in rabbit, Anti-B7 homolog 3 antibody produced in rabbit, Anti-B7-H3 antibody produced in rabbit, Anti-CD276 antigen precursor antibody produced in rabbit, Anti-Costimulatory molecule antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique