
Anilofos

Synonym(s):
S-[2-(4-Chloro-N-isopropylanilino)-2-oxoethyl]-O,O′-dimethyl dithiophosphate
Empirical Formula (Hill Notation):
C13H19ClNO3PS2
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2