
α-Methylserotonin maleate salt

Synonym(s):
α-Methyl-5-hydroxytryptamine maleate salt, (±)-3-(2-Aminopropyl)indol-5-ol maleate salt
Empirical Formula (Hill Notation):
C11H14N2O · C4H4O4
CAS Number:
Molecular Weight:
306.31
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing