Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
AmberChrom
™
50WX4 Ion Exchange Resin
Linear Formula:
(C
10
H
12
· C
10
H
10
· C
8
H
8
)
x
CAS Number:
69011-20-7
Products
Product Category
resins (3)
Physical Form
Separation Technique
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
217484
AmberChrom
™
50WX4 Ion Exchange Resin,
hydrogen form, 200-400 mesh, cationic
Expand
View Pricing
422096
AmberChrom
™
50WX4 Ion Exchange Resin,
hydrogen form, 100-200 mesh
Expand
View Pricing
428663
AmberChrom
™
50WX4 Ion Exchange Resin,
hydrogen form, 50-100 mesh
Expand
View Pricing