
Adapalene Related Compound A

Synonym(s):
Methyl 6-bromo-2-naphthoate
Linear Formula:
BrC10H6CO2CH3
CAS Number:
Molecular Weight:
265.10
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2