
Acrolein-2,4-DNPH

Synonym(s):
Acrolein-2,4-dinitrophenylhydrazone
Empirical Formula (Hill Notation):
C9H8N4O4
CAS Number:
Molecular Weight:
236.18
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing