
6-Nitrochrysene

Empirical Formula (Hill Notation):
C18H11NO2
CAS Number:
Molecular Weight:
273.29
Products
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing