
5-Hexen-2-one

Synonym(s):
Allylacetone
Linear Formula:
CH3COCH2CH2CH=CH2
CAS Number:
Molecular Weight:
98.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-2 of 2
Compare
Product Number
Description
Pricing