![5-Bromo-1,2,3-trifluorobenzene 99%](/deepweb/assets/sigmaaldrich/product/structures/130/880/2eaf4af0-6132-4e1b-9c05-edcb6c35386c/640/2eaf4af0-6132-4e1b-9c05-edcb6c35386c.png)
5-Bromo-1,2,3-trifluorobenzene
![5-Bromo-1,2,3-trifluorobenzene 99%](/deepweb/assets/sigmaaldrich/product/structures/130/880/2eaf4af0-6132-4e1b-9c05-edcb6c35386c/640/2eaf4af0-6132-4e1b-9c05-edcb6c35386c.png)
Synonym(s):
1-Bromo-3,4,5-trifluorobenzene
Linear Formula:
BrC6H2F3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing