2-Imidazolidone-4-carboxylic acid
Synonym(s):
2-Oxo-4-imidazolidinecarboxylic acid
Empirical Formula (Hill Notation):
C4H6N2O3
CAS Number:
Molecular Weight:
130.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1