
2-Amino-5-phenyl-1,3,4-oxadiazole

Empirical Formula (Hill Notation):
C8H7N3O
CAS Number:
Molecular Weight:
161.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing