Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-Thioglycerol
Synonym(s):
α-Monothioglycerol, α-Thioglycerol, 3-Mercapto-1,2-propanediol
Linear Formula:
HSCH
2
CH(OH)CH
2
OH
CAS Number:
96-27-5
Molecular Weight:
108.16
EC Number:
202-495-0
Products
Product Category
glycerols (3)
lipids (3)
Brand
SAFC (1)
Sigma-Aldrich (4)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
M6145
1-Thioglycerol,
liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Expand
View Pricing
M1753
1-Thioglycerol,
≥97%
Expand
View Pricing
108704
1-Thioglycerol,
EMPROVE
®
ESSENTIAL, NF
Expand
View Pricing
88640
1-Thioglycerol,
≥99.0% (GC)
Expand
View Pricing
M2172
1-Thioglycerol,
BioXtra, ≥97%
Expand
View Pricing