Skip to Content
Merck
1-Naphthol SAJ special grade, ≥99.0%

1-Naphthol

1-Naphthol SAJ special grade, ≥99.0%
Synonym(s):
α-Naphthol, 1-Hydroxynaphthalene, 1-Naphthol
Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group