Skip to Content
Merck
1-Fluoropyridinium triflate 99%

1-Fluoropyridinium triflate

1-Fluoropyridinium triflate 99%
Synonym(s):
1-Fluoropyridinium trifluoromethanesulfonate
Empirical Formula (Hill Notation):
C6H5F4NO3S
CAS Number:
Molecular Weight:
247.17

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing