
1-Ethyl-1-methylpyrrolidinium tetrafluoroborate

Empirical Formula (Hill Notation):
C7H16BF4N
CAS Number:
Molecular Weight:
201.01
Products
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing