![1,3-Dibromo-5,5-dimethylhydantoin 98%](/deepweb/assets/sigmaaldrich/product/structures/146/897/c4c0fe79-9ae7-481e-aa45-4d21c9e59ec4/640/c4c0fe79-9ae7-481e-aa45-4d21c9e59ec4.png)
1,3-Dibromo-5,5-dimethylhydantoin
![1,3-Dibromo-5,5-dimethylhydantoin 98%](/deepweb/assets/sigmaaldrich/product/structures/146/897/c4c0fe79-9ae7-481e-aa45-4d21c9e59ec4/640/c4c0fe79-9ae7-481e-aa45-4d21c9e59ec4.png)
Synonym(s):
Dibromantin, 1,3-Dibromo-5,5-dimethylhydantoin
Empirical Formula (Hill Notation):
C5H6Br2N2O2
CAS Number:
Molecular Weight:
285.92
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing