Skip to Content
Merck
All Photos(1)

Documents

1613407

USP

Sodium acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sodium acetate

form

solid

autoignition temp.

1112 °F

manufacturer/tradename

USP

pH

8-9.5

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabina Lukovac et al.
mBio, 5(4) (2014-08-15)
The gut microbiota is essential for numerous aspects of human health. However, the underlying mechanisms of many host-microbiota interactions remain unclear. The aim of this study was to characterize effects of the microbiota on host epithelium using a novel ex
Carol L Ladner-Keay et al.
PloS one, 9(6), e98753-e98753 (2014-06-04)
The formation of β-sheet rich prion oligomers and fibrils from native prion protein (PrP) is thought to be a key step in the development of prion diseases. Many methods are available to convert recombinant prion protein into β-sheet rich fibrils
Sam Joshva Baskar Jesudasan et al.
PloS one, 9(6), e99443-e99443 (2014-06-11)
Microglia are the primary immune cells of the central nervous system (CNS). Membrane bound sensors on their processes monitor the extracellular environment and respond to perturbations of the CNS such as injury or infection. Once activated, microglia play a crucial
Mari Endo et al.
Journal of gastroenterology, 49(6), 1026-1039 (2013-07-13)
Daikenchuto (DKT), a gastrointestinal prokinetic Japanese herbal medicine, is prescribed for patients with postoperative ileus (POI) and adhesive bowel obstruction following abdominal surgery. Several mechanisms for the amelioration of POI by DKT have been suggested; however, it has remained unclear
Cristina Basilico et al.
The Journal of clinical investigation, 124(7), 3172-3186 (2014-05-29)
Activation of MET by HGF plays a key role in tumor progression. Using a recently developed llama platform that generates human-like immunoglobulins, we selected 68 different antibodies that compete with HGF for binding to MET. HGF-competing antibodies recognized 4 distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service