Skip to Content
Merck
All Photos(1)

Key Documents

1524012

USP

Phenobarbital Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6-Amino-5-ethyl-2-imino-5-phenylpyrimidin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O
CAS Number:
Molecular Weight:
230.27
UNSPSC Code:
41116107
NACRES:
NA.24

API family

phenobarbital

packaging

pkg of 30 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecules)

format

neat

storage temp.

2-8°C

InChI

1S/C12H14N4O/c1-2-12(8-6-4-3-5-7-8)9(13)15-11(14)16-10(12)17/h3-7H,2H2,1H3,(H4,13,14,15,16,17)

InChI key

ZKPNDYQXPJGNCR-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Phenobarbital Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service