Skip to Content
Merck
All Photos(1)

Key Documents

69705

Supelco

2-(2H-Benzotriazol-2-yl)-4-methyl-6-(2-propenyl)phenol

analytical standard

Synonym(s):

2-(3-Allyl-2-hydroxy-5-methylphenyl)-2H-benzotriazole, 2-Allyl-6-(2H-benzotriazol-2-yl)-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O
CAS Number:
Molecular Weight:
265.31
MDL number:

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

98-101 °C (lit.)

transition temp

transition temp >270 °C

solubility

H2O: insoluble

density

1.3 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Cc1cc(CC=C)c(O)c(c1)-n2nc3ccccc3n2

InChI

1S/C16H15N3O/c1-3-6-12-9-11(2)10-15(16(12)20)19-17-13-7-4-5-8-14(13)18-19/h3-5,7-10,20H,1,6H2,2H3

InChI key

YKONWVIRECCMQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Covalently bondable UV light stabilizer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service