Key Documents
58442
Supelco
SSI LO-Pulse Damper
without cabinet
About This Item
UNSPSC Code:
41115854
description
316 stainless steel
cabinet no
feature
without cabinet
General description
A pulse damper controls pump pulsations for a more stable baseline. The LO-Pulse damper is a patented unit compatible
with single piston reciprocating HPLC pumps (Altex 110A, Eldex pumps, LDC Mini-Pump VS, SSI Models 200 and 300,
etc.). At pressures from 500 psi to 6,000 psi (35-420 kg/cm2), it improves precision of quantitative analyses and detection limits for trace sample components. Fittings and instructions included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
LO-Pulse is a trademark of Scientific Systems, Inc.
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.