Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1293

Sigma-Aldrich

Anti-CACNA1C Antibody, clone 1F20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F20, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 248.98 kDa
observed mol wt ~250 kDa

species reactivity

human, rat

species reactivity (predicted by homology)

monkey, bovine, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1F20 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Calcium Channel- 1C Subunit (CACNA1C). It targets an epitope within 17 amino acids from the 7th cytoplasmic domain within the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the 7th cytoplasmic domain of human Voltage-dependent L-type calcium channel subunit a-1C (CACNA1C).

Application

Quality Control Testing

Evaluated by Western Blotting in Human Heart tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CACNA1C in Human Heart tissue lysate.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CACNA1C in L6 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CACNA1C in human heart and human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CACNA1C, clone 1F20 ZooMAb®, Cat. No. ZRB1293, is a recombinant Rabbit monoclonal antibody that specifically targets CACNA1C and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Voltage-dependent L-type calcium channel subunit alpha-1C (UniProt: Q13936; also known as Calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle Voltage-gated calcium channel subunit alpha Cav1.2, CACNA1C) is encoded by the CACNA1C (also known as CACH2, CACN2, CACNl1A1, CCHL1A1) gene (Gene ID: 775) in human. CACNA1C is a multi-pass membrane protein that serves as a pore-forming alpha-1C subunit of the voltage-gated calcium channel that gives rise to L-type calcium currents. It colocalizes with ryanodine receptors in distinct clusters at the junctional membrane, where the sarcolemma and the sarcoplasmic reticulum are in close contact. It mediates the influx of calcium ions into the cytoplasm, and thereby triggers calcium release from the sarcoplasm. It is also reported to play a role in excitation-contraction coupling in the heart and is required for normal heart development and normal regulation of cardiac rhythm. Its phosphorylation by protein kinase A at seine 1981 is shown to activate the channel. CACNA1C is also shown to be essential for normal contraction of smooth muscle cells in blood vessels that is required for normal blood pressure regulation. Its activity can be inhibited by treatment with nifedipine and dihydropyrdines. Mutations in CACNa1C gene are known to cause long QT syndrome that is characterized by a prolonged QT interval on the ECG and polymorphic ventricular arrhythmias. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service