Skip to Content
Merck
All Photos(1)

Key Documents

SML2389

Sigma-Aldrich

AP39

≥95% (HPLC)

Synonym(s):

(10-Oxo-10-(4-(3-thioxo-3H-1,2-dithiol-5-yl)phenoxy)decyl)triphenylphosphonium bromide, AP 39, AP-39

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H38BrO2PS3
CAS Number:
Molecular Weight:
721.77
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to dark orange

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(OC1=CC=C(C2=CC(SS2)=S)C=C1)CCCCCCCCC[P+](C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5.[Br-]

InChI

1S/C37H38O2PS3.BrH/c38-36(39-31-26-24-30(25-27-31)35-29-37(41)43-42-35)23-15-4-2-1-3-5-16-28-40(32-17-9-6-10-18-32,33-19-11-7-12-20-33)34-21-13-8-14-22-34;/h6-14,17-22,24-27,29H,1-5,15-16,23,28H2;1H/q+1;/p-1

InChI key

PUEBFERUDPTIDR-UHFFFAOYSA-M

Biochem/physiol Actions

AP39 is a anethole dithiolethione (ADT-OH)-derivatized hydrogen sulfide (H2S) donor with a lipophilic & positively charged triphenylphosphonium (TP) moiety for mitochondrially targeted H2S release. AP39 stimulates mitochondrial electron transport and cellular bioenergetic function, protecting against mitochondrial DNA damage and cell death upon oxidative stress induction (effective conc. 30–100 nM; bEnd.3 cells). AP39 in vivo efficacy (0.1-1 μmol/kg i.v.; 0.1-0.3 mg/kg i.p.) is demonstrated in varous rodent models, including neurological damage post cardiac arrest, renal injury post ischemia-reperfusion, burn-caused organ damage, as well as brain atrophy in APP/PS1 mice.
Mitochondrially targeted hydrogen sulfide (H2S) donor that protects against mitochondrial stress-induced cellular and tissue damage in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service