Skip to Content
Merck
All Photos(2)

Key Documents

SAE0095

Sigma-Aldrich

Agrin Human

Recombinant, cell culture tested, expressed in HEK 293 cells

Synonym(s):

AGRN, Proteoglycan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

attachment assay, measured by the ability of the immobilized agrin to support the adhesion of PC12 cells

technique(s)

cell culture | mammalian: suitable

suitability

endotoxin tested

shipped in

ambient

storage temp.

−20°C

General description

Agrin C-terminal human recombinant is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of 87 kDa (amino acids Ala1260-Pro2045, with an N-terminal His tag). The DTT-reduced protein migrates as a ∼100 kDa polypeptide on SDS-PAGE due to glycosylation. This protein is manufactured in human cells, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports higher bioactivity of the protein.

Biochem/physiol Actions

Agrin is a large extracellular heparan sulfate proteoglycan that is involved in the formation of the neuromuscular junction.
Agrin triggers the aggregation of acetylcholine receptors via the muscle-specific kinase (MuSK) and the low-density lipoprotein receptor-related protein 4 (Lrp4) receptor complex.
Agrin is required for the full regenerative capacity of neonatal mouse hearts.
Recombinant agrin has been shown in vitro to promote cardiomyocyte proliferation, as studied in mouse-induced and human-induced pluripotent stem cells. Recombinant agrin has been shown in vivo to promote cardiac regeneration, after induction of myocardial infarction in adult mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiu-Qing Fu et al.
Cell discovery, 6, 9-9 (2020-03-07)
During the development of mammalian neuromuscular junction (NMJ), the original supernumerary axon inputs are gradually eliminated, finally leaving each muscle fiber innervated by a single axon terminal. However, the molecular cues that mediate the elimination of redundant axon inputs remain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service