Skip to Content
Merck
All Photos(1)

Key Documents

RAB0349

Sigma-Aldrich

Phospho-Erk1 (pThr202 / pTyr204 + Erk2 (pTyr185/187) and pan-Erk1 / 2 ELISA Kit

for detection of phospho-erk1 and phospho-erk2 and pan-erk1/2 in human, mouse or rat cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat, mouse, human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAPK3(5595)

General description

The Phospho-Erk1(pThr202 / pTyr204) + Erk2(pTyr185/187) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.

Immunogen

ERK1 (pThr202) synthetic phosphopeptide, Recombinant ERK1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service