Skip to Content
Merck
All Photos(1)

Key Documents

N6877

Sigma-Aldrich

L-Norleucine

≥98% (TLC)

Synonym(s):

(S)-(+)-2-Aminohexanoic acid, (S)-2-Aminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721750
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥98% (TLC)

form

powder

color

white

mp

>300 °C (lit.)

application(s)

detection

SMILES string

CCCC[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m0/s1

InChI key

LRQKBLKVPFOOQJ-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • l-norleucine on high glucose-induced insulin sensitivity and mitochondrial function in skeletal muscle cells. This study explores the beneficial effects of L-Norleucine on insulin sensitivity and mitochondrial function, underlining its potential in metabolic pathway research and its application in managing diabetes through targeted amino acid therapy (Ding et al., 2024).

Biochem/physiol Actions

L-Norleucine is a synthetic amino acid commonly used as an internal standard.

Other Notes

Non-essential amino acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luana Bontempo et al.
Food chemistry, 318, 126426-126426 (2020-03-07)
Until now, there has been a lack of analytical methods that can reliably verify the authenticity of organically grown plants and derived organic food products. In this study, stable isotope ratio analysis of hydrogen (H, δ2H), carbon (C, δ13C), nitrogen
Alborz Mahdavi et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 433-438 (2013-12-19)
Pathogenic microbes have evolved complex secretion systems to deliver virulence factors into host cells. Identification of these factors is critical for understanding the infection process. We report a powerful and versatile approach to the selective labeling and identification of secreted
Troy Cellmer et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(15), 6103-6108 (2011-03-29)
Determining the rate of forming the truly folded conformation of ultrafast folding proteins is an important issue for both experiments and simulations. The double-norleucine mutant of the 35-residue villin subdomain is the focus of recent computer simulations with atomistic molecular
Silvia Lisa et al.
Cellular and molecular life sciences : CMLS, 67(16), 2825-2838 (2010-05-11)
The conversion of the cellular prion protein (PrP(C)) into its disease-associated form (PrP(Sc)) involves a major conformational change and the accumulation of sulfoxidized methionines. Computational and synthetic approaches have shown that this change in the polarity of M206 and M213
Haiyan Wei et al.
The journal of physical chemistry. B, 114(36), 11820-11826 (2010-08-24)
Following the studies of urea denaturation of β-hairpins using molecular dynamics, in this paper, molecular dynamics simulations of two peptides, a 35 residue three helix bundle villin headpiece protein HP-35 and its doubly norleucine-substituent mutant (Lys24Nle/Lys29Nle) HP-35 NleNle, were undertaken

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service