Skip to Content
Merck
All Photos(7)

Documents

HPA016820

Sigma-Aldrich

Anti-PCNT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Kendrin, Anti-Pericentrin, Anti-Pericentrin B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MEDLQNQFQKELAEQRAELEKIFQDKNQAERALRNLESHHQAAIEKLREDLQSEHGRCLEDLEFKFKESEKEKQLELENLQASYEDLKAQSQEEIRRLWSQLDSARTSRQELSELHEQLLARTSRVEDLEQLKQREKTQHESELE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCNT(5116)

General description

PCNT (pericentrin) is a centrosomal protein mapped on chromosome 21q22.3. It is localized in the centrosome as an integral component of the pericentriolar material.

Immunogen

Pericentrin recombinant protein epitope signature tag (PrEST)

Application

Anti-PCNT antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PCNT (pericentrin) is associated with various centrosomal activities. It forms a functional complex with pericentriolar material 1 (PCM1) in cells, which is responsible for centrosome function. In association with DISC1 (Disrupted-In-Schizophrenia 1), PCNT performs a vital role in the microtubule network assembly. During cytoskeleton assembly, it binds to the microtubule nucleation component, γ-tubulin, and plays a crucial role in the cell-cycle progression by directing a proper spindle organization. It has been observed that low level of PCNT alters the arrangement of mitotic spindles and leads missegregation of chromosomes. Mutation in PCNt gene causes two autosomal recessive disorders named Seckel syndrome and Majewski osteodysplastic primordial dwarfism type II (MOPD II).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73892

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Pasdeloup et al.
Journal of virology, 87(12), 7102-7112 (2013-04-19)
Efficient intracellular transport of the capsid of alphaherpesviruses, such as herpes simplex virus 1 (HSV-1), is known to be dependent upon the microtubule (MT) network. Typically, the MT network radiates from an MT-organizing center (MTOC), which is, in most cases
Anita Rauch et al.
Science (New York, N.Y.), 319(5864), 816-819 (2008-01-05)
Fundamental processes influencing human growth can be revealed by studying extreme short stature. Using genetic linkage analysis, we find that biallelic loss-of-function mutations in the centrosomal pericentrin (PCNT) gene on chromosome 21q22.3 cause microcephalic osteodysplastic primordial dwarfism type II (MOPD
Maria Piane et al.
American journal of medical genetics. Part A, 149A(11), 2452-2456 (2009-10-20)
We report on a 3-year-old boy with prenatal onset of proportionate dwarfism, postnatal severe microcephaly, high forehead with receded hairline, sparse scalp hair, beaked nose, mild retrognathia and hypotonia diagnosed at birth as Seckel syndrome. At age 3 years, he
Shuk-Mei Ho et al.
Endocrine-related cancer, 24(2), 83-96 (2016-12-22)
Humans are increasingly exposed to structural analogues of bisphenol A (BPA), as BPA is being replaced by these compounds in BPA-free consumer products. We have previously shown that chronic and developmental exposure to BPA is associated with increased prostate cancer
Q Li et al.
Journal of cell science, 114(Pt 4), 797-809 (2001-02-15)
The centrosome is responsible for nucleating microtubules and performing other cellular roles. To define the organization of the centrosome more completely, a human anti-centrosome serum was used to screen a human cDNA library, and a cDNA encoding a >350 kDa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service