Skip to Content
Merck
All Photos(1)

Documents

GEPA15102

Sigma-Aldrich

Cy®5 Mono NHS Ester

Cytiva PA15102, pack of 50 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

packaging

pack of 50 mg

manufacturer/tradename

Cytiva PA15102

color

blue
Blue

storage temp.

2-8°C

General description

The packs contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

Application

For labelling via amine groups. CyDyes are suitable for use in applications such as fluorescence resonance energy transfer (FRET), time resolved- fluorescence resonance energy transfer (TR-FRET), and fluorescence polarization (FP).

Features and Benefits

  • Targeted and specific labeling of amine residues on antibodies, peptides or oligonucleotides.
  • Bulk packs available for greatest economy.
  • Example protocols included with each dye pack.

Components

foil packs, each containing 1, 5, 10, 25 or 50 mg dried dye.

Caution

Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact LSlicensing@cytiva.com.

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy is a registered trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service