Skip to Content
Merck
All Photos(1)

Key Documents

CLS3415

Corning® Transwell® 24 well plates

3.0 μm pore size, Tissue Culture (TC)-treated surface round wells, clear bottom, clear wells, flat bottom wells, clear polystyrene (plate or dish), polycarbonate membrane, sterile, case of 48, pack of 12

Synonym(s):

cell culture inserts, corning plates, tissue culture plate, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

product name

Corning® Transwell® polycarbonate membrane cell culture inserts, 6.5 mm Transwell with 3.0 μm pore polycarbonate membrane insert, TC-treated, w/ lid, sterile, 48/cs

material

clear bottom
clear polystyrene (plate or dish)
clear wells
flat bottom wells
polycarbonate membrane
round wells

sterility

sterile

feature

lid
skirt

packaging

pack of 12
case of 48

manufacturer/tradename

Corning 3415

technique(s)

cell culture | mammalian: suitable

insert working volume

0.1 mL

membrane diam.

6.5 mm

size

24 wells

surface area

0.33 cm2 , cell growth area

well working volume

0.6 mL

color

translucent

pore size

3.0 μm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • 10μm thick translucent polycarbonate membrane
  • Treated for optimal cell attachment
  • Packaged 12 inserts in a 24 well plate, 4 plates per case
  • Membrane must be stained for cell visibility
  • Sterilized by gamma radiation
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines. They feature a 10 μm thin transparent polycarbonate membrane available in a variety of pore sizes. Transwell are treated for optimal cell attachment. Cells must be stained for cell visibility. The PC membrane is compatible with most organic fixatives and stains.

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service