Skip to Content
Merck
All Photos(1)

Documents

79454

Sigma-Aldrich

Ampliflu Red Western Blot Kit

for fluorescent detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 50 applications

fluorescence

λex 571 nm; λem 585 nm

storage temp.

2-8°C

General description

<p>find more information here</p>

Application

The Ampliflu Red Western Blot Kit provides an alternative to the classical chemiluminescence detection. When working with Ampliflu Red, standard protocols for the immunoblotting-procedure can be used until the detection step. No extra time for signal accumulation is necessary as it is for chemiluminescent signals. The use of Ampliflu Red is suitable for protein quantities between 1 ng and 1 μg per band of protein.

Principle

The kit contains the peroxidase substrate Ampliflu Red that is enzymatically converted to the highly fluorescent compound, resorufin. Fluorescent imaging is then performed on the immunoblot.

Analysis Note

Sensitivity: Limit of detection is 1 ng/band, but may vary depending on the antibodies used.

Legal Information

Ampliflu is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC) 3 mL

Kit Components Also Available Separately

Product No.
Description
SDS

  • 90101Ampliflu Red, for fluorescence, ≥98.0% (HPLC) 5 vial(s)SDS

  • 79382Phosphate buffered saline, tablet 5 tabletsSDS

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service