Skip to Content
Merck
All Photos(3)

Key Documents

A2140

Sigma-Aldrich

Ammonium aluminum sulfate dodecahydrate

ReagentPlus®, ≥99% (titration)

Synonym(s):

Aluminum ammonium sulfate dodecahydrate, Ammonium alum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlNH4(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
453.33
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99% (titration)
form:
powder or crystals

Quality Level

product line

ReagentPlus®

Assay

≥99% (titration)

form

powder or crystals

pH

2.6 (25 °C, 100 g/L)

mp

93.5 °C (lit.)

SMILES string

[Al+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H][N+]([H])([H])[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

WZUKKIPWIPZMAS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium aluminum sulfate dodecahydrate, commonly called as ammonium alum, is widely used in the water purification, cosmetic formulation, fertilizers and food preservation. Its thermal decomposition has been studied. It has high latent heat of 251kJkg-1 with a potential use in the heating system. Its utility as a flame-retardant for cotton fabric has been studied.

Application

Ammonium aluminum sulfate dodecahydrate may be used in the preparation of alum (Al(OH)3) gel and monodispersed yttrium aluminum garnet (YAG) powder.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of monodispersed spherical yttrium aluminum garnet (YAG) powders by a homogeneous precipitation method.
Xu X, et al.
Journal of the American Ceramic Society. American Ceramic Society, 95(12), 3821-3826 (2012)
Preventive and curative glycoside kaempferol treatments attenuate the TH2-driven allergic airway disease.
Medeiros KCP, et al.
International Immunopharmacology, 9(13), 1540-1548 (2009)
Thermogravimetry of deposited ammonium aluminum sulfate dodecahydrate used as flame-retardant for cotton fabrics.
Mostashari SM and Mostashari SZ.
Cellulose Chemistry and Technology, 43(9), 455-455 (2009)
Flow and heat transfer characteristics of ammonium alum hydrate slurries.
Suzuki H, et al.
International Journal of Refrigeration, 36(1), 81-87 (2013)
Thermal characterization of ammonium alum.
Lopez-Beceiro J, et al.
Journal of Thermal Analysis and Calorimetry, 104(1), 127-130 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service