Skip to Content
Merck
All Photos(1)

Key Documents

Y0001404

Vigabatrin impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

γ-Aminobutyric acid, 3-Carboxypropylamine, 4-Aminobutanoic acid, GABA, Piperidic acid, Piperidinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
906818
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vigabatrin

manufacturer/tradename

EDQM

mp

195 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NCCCC(O)=O

InChI

1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)

InChI key

BTCSSZJGUNDROE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vigabatrin impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Major inhibitory neurotransmitter in brain; GABAA and GABAB receptor agonist; increases Cl conductance.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jesse Jackson et al.
Nature neuroscience, 17(10), 1362-1370 (2014-09-01)
Activity flow through the hippocampus is thought to arise exclusively from unidirectional excitatory synaptic signaling from CA3 to CA1 to the subiculum. Theta rhythms are important for hippocampal synchronization during episodic memory processing; thus, it is assumed that theta rhythms
Hussein A N Al-Wadei et al.
Molecular nutrition & food research, 55(12), 1745-1758 (2011-08-02)
GABA is a bioactive constituent of fruits, vegetables, cereals and is believed to play a role in defense against stress in plants. In animals, it acts as an inhibitory neurotransmitter in brain while also expressed in non-neuronal cells. Studies have
Melissa A Farmer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(17), 5747-5753 (2014-04-25)
Chronic pain is often associated with sexual dysfunction, suggesting that pain can reduce libido. We find that inflammatory pain reduces sexual motivation, measured via mounting behavior and/or proximity in a paced mating paradigm, in female but not male laboratory mice.
A D Stan et al.
Molecular psychiatry, 20(4), 433-439 (2014-06-11)
Hippocampal dysfunction in schizophrenia is widely acknowledged, yet the mechanism of such dysfunction remains debated. In this study we investigate the excitatory and inhibitory hippocampal neurotransmission using two complementary methodologies, proton magnetic resonance spectroscopy (MRS) and tissue biochemistry, sampling individuals
Amanda Peltier et al.
BMJ (Clinical research ed.), 348, g1799-g1799 (2014-05-08)
Diabetes is a worldwide epidemic, and associated neuropathy is its most costly and disabling complication. Given the rising prevalence of painful diabetic neuropathy, it is increasingly important that we understand the best ways to diagnose and treat this condition. Diagnostic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service