Skip to Content
Merck
All Photos(1)

Documents

SMB01383

Sigma-Aldrich

PhIP

≥95% (HPLC)

Synonym(s):

2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N4
CAS Number:
Molecular Weight:
224.26
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.28

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

shipped in

2-8°C

storage temp.

2-8°C

SMILES string

NC1=NC2=NC=C(C3=CC=CC=C3)C=C2N1C

Looking for similar products? Visit Product Comparison Guide

General description

PhIP (2-Amino-1-methyl-6-phenylimidazo(4,5-b)pyridine) is a dietary genotoxic carcinogen formed in heat processed meat and fish via the Maillard reaction. PhIP is a primary metabolite activated by CYP1A2 and N-acetyltransferase 1 or sulfotransferase to DNA reactive metabolite.

Biochem/physiol Actions

Carcinogen developed in grilled meats. Induces breast and colon cancer in rats.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yueliang Zhao et al.
Journal of agricultural and food chemistry, 69(46), 13933-13941 (2021-11-11)
The effects of the deacetylation degree (DD) of chitosan on heterocyclic aromatic amine formation were investigated in chemical models and beef patties. The results in model systems showed that at lower addition levels (10 mg), chitosan with 85% DD showed
Robbert Ijsselsteijn et al.
Carcinogenesis, 43(2), 160-169 (2021-12-18)
The prevalent cancer predisposition Lynch syndrome (LS, OMIM #120435) is caused by an inherited heterozygous defect in any of the four core DNA mismatch repair (MMR) genes MSH2, MSH6, MLH1 or PMS2. MMR repairs errors by the replicative DNA polymerases
Iftikhar Ali Khan et al.
Meat science, 184, 108693-108693 (2021-11-15)
At this point in time, the evidence of a link between well-done meat intake and the incidence of cancer is stronger than it was 20 years ago. Several cohort and case-control studies have confirmed this evidence, and have shown a
Shambhunath Choudhary et al.
Carcinogenesis, 33(4), 876-885 (2012-02-07)
More than 85% of breast cancers are sporadic and attributable to long-term exposure to environmental carcinogens, such as those in the diet, through a multistep disease process progressing from non-cancerous to premalignant and malignant stages. The chemical carcinogen 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service