Skip to Content
Merck
All Photos(1)

Documents

PHR1433

Supelco

Ascorbyl Glucoside

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ascorbyl Glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O11
CAS Number:
Molecular Weight:
338.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

ascorbyl glucoside

form

solid

CofA

current certificate can be downloaded

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(C1=O)O[C@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO

InChI

1S/C12H18O11/c13-1-3(15)9-8(19)10(11(20)22-9)23-12-7(18)6(17)5(16)4(2-14)21-12/h3-7,9,12-19H,1-2H2/t3-,4+,5+,6-,7+,9+,12+/m0/s1

InChI key

MLSJBGYKDYSOAE-DCWMUDTNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ascorbyl Glucoside is used to protect skin from exposure to UV rays.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2741 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basem Kanawati et al.
Rapid communications in mass spectrometry : RCM, 25(6), 806-814 (2011-02-22)
L-Ascorbic acid and two distinct anomers, namely the α-D-glucopyranosyl and β-D-glucopyranosyl-(1→2)-L-ascorbic acid (stereoisomers), were studied within the scope of collision-induced dissociation (CID) experiments, performed by linear ion acceleration and collision with argon atoms inside a hexapole quadrupole hexapole ion beam
Akihiro Tai et al.
Carbohydrate research, 345(12), 1658-1662 (2010-06-23)
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid (AA-2G) laurate was synthesized from AA-2G and vinyl laurate with a protease from Bacillus subtilis in N,N-dimethylformamide (DMF) with low water content. Addition of water to DMF dramatically enhanced monoacyl AA-2G synthesis. Maximum synthetic activity was observed when
Yuko Kinashi et al.
Mutation research, 695(1-2), 69-74 (2009-12-17)
Evidence from in vivo studies suggests there are enhanced radiation effects in abscopal regions after local head gamma ray irradiation. Splenocyte apoptosis and T lymphocyte micronuclei were induced at higher rates than what would be estimated given the dose at
Mutsuko Taniguchi et al.
European journal of pharmacology, 674(2-3), 126-131 (2011-11-29)
A stable ascorbic acid derivative, 2-O-α-glucopyranosyl-l-ascorbic acid (AA-2G), was evaluated and compared with ascorbic acid for its protective effect against cellular damage and senescence induced by hydrogen peroxide (H(2)O(2)). Pretreatment with AA-2G for 72 h promoted the proliferation of normal
Kenji Ichiyama et al.
Immunology letters, 122(2), 219-226 (2009-02-10)
The stable ascorbic acid derivative 2-O-alpha-D-glucopyranosyl-L-ascorbic acid (AA-2G) was used to investigate the role of ascorbic acid (AA) in B cell differentiation in vitro. AA-2G is stable in a solution unlike AA but is hydrolyzed by cellular alpha-glucosidase to release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service